Idex Port Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Port Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA
Idex | Mfr: P-314| Item: 0201834
Available in various thread sizes and materials
₹770.00INR / Each
₹770.00INR / Each
Buy It Together
Current Item
Idex Port Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA
Idex Port Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA
0201834
₹770.00INR / Each
+ =
Total Price 1410.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • For M6 flat-bottom port
  • Black ETFE construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Year Warranty
Specifications
  • MaterialETFE
  • ColorBlack
  • Compatible WithM6 flat-bottom port
  • DescriptionPort Plug, Standard Knurl, Black ETFE, M6 Flat-Bottom; 1/EA