Idex Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-309| Item: 0201377
Available in various thread sizes and materials
₹490.00INR / Each
₹490.00INR / Each
Buy It Together
Current Item
Idex Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA
Idex Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA
0201377
₹490.00INR / Each
+ =
Total Price 1130.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • For 1/4-28 flat-bottom port
  • Black acetal construction
  • 1 month warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Month Warranty
Specifications
  • MaterialDelrin® acetal
  • ColorBlack
  • Compatible With1/4-28 flat-bottom port
  • DescriptionP-309 Port Plug, Standard Knurl, Black Acetal, 1/4-28 Flat-Bottom; 1/EA