Idex Port Plug, Micro Headless, PEEK, Green, 6-40 Flat-Bottom
Representative image only.
Idex Port Plug, Micro Headless, PEEK, Green, 6-40 Flat-Bottom
Idex | Mfr: P-558| Item: 0201846
Close unused ports to safely store sample
₹1,600.00INR / Each
₹1,600.00INR / Each
Product Description
Key Features
  • Constructed of chemically inert PEEK plastic
  • For 6-40 flat-bottom port
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorGreen
  • Compatible With6-40 flat-bottom port
  • DescriptionPort Plug, Micro Headless, Green PEEK, 6-40 Flat-Bottom; 1/EA