Idex PEEK Needle Port for Sample Injection Valves
Representative image only.
Idex PEEK Needle Port for Sample Injection Valves
Idex | Mfr: 9013| Item: 0203206
Prevent samples from veering away from the flow path
₹13,700.00INR / Each
₹13,700.00INR / Each
Product Description
Key Features
  • For Use with Any Standard High Pressure Injection Valve
  • For Use with Any Standard High Pressure Injection Valve
  • 1 year warranty
More About this Item
These adapters are made so the least amount of sample is wasted.
A. Injection Port Adapters eliminate the potential for additional swept volume to the fluid pathway since the needle butts directly against the connecting tubing. They can be bulkhead mounted or mounted with the insulated mounting bracket assembly 02032-11.
B. Suction Needle Adapters allows for suction-loading of a valve for metal-free loading of a sample loop.