Idex O-Rings for MLP and MXX Valves, 1/16" OD Tubing; 25/PK
Representative image only.
Idex O-Rings for MLP and MXX Valves, 1/16" OD Tubing; 25/PK
Idex | Mfr: 7770-124| Item: 0202526
Optimize performance of your Idex® valve
₹55,100.00INR / Pkg of 25
₹55,100.00INR / Pkg of 25
Product Description
Key Features
  • Choose from replacement fittings and o-rings to extend the life of your valves
  • For MLP and MXX valves
  • 1 year warranty
More About this Item
Replacement ferrules and O-rings for valves offer a tight fit for 1/16" and 1/8" OD tubing.
1 Year Warranty
Specifications
  • Compatible WithMLP and MXX valve
  • DescriptionIdex O-Rings for MLP and MXX Valves, 1/16" OD Tubing; 25/PK