Idex NanoTight™ Fitting, Headless, Short, Natural PEEK/ETFE, Max 0.063" Hole, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex NanoTight™ Fitting, Headless, Short, Natural PEEK/ETFE, Max 0.063" Hole, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: F-333NX| Item: 0201349
Keep you operation running smoothly by keeping replacement parts handy
₹22,500.00INR / Pkg of 10
₹22,500.00INR / Pkg of 10
Product Description
Key Features
  • Optimize performance of your Idex® NanoPort
  • 1/32" OD x 150 µm ID x 50 cm L
  • 1 month warranty
  • 1 month warranty
More About this Item
Choose from a wide range of replacement parts including ferrules, gaskets, and more.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK/ETFE
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(M) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)4000
  • DescriptionNanoTight™ Fitting, Headless, Short, Natural PEEK/ETFE, Max 0.063" Hole, 1/16" OD Tubing, 10-32 Coned; 10/PK