Idex MicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA
Representative image only.
Idex MicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA
Idex | Mfr: P-554| Item: 0202433
Available in various thread sizes and materials
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Idex MicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA
Idex MicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA
0202433
₹2,400.00INR / Each
+ =
Total Price 3040.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • Achieve True ZDV connections
  • Max pressure 1000 psi (345 bar)
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
Specifications
  • MaterialDelrin® acetal
  • ColorWhite
  • Compatible With10-32 Coned Port
  • DescriptionMicroTight Guage Plug, White Acetal, 10-32 Coned; 1/EA