Idex MicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA
Representative image only.
Idex MicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA
Idex | Mfr: P-553| Item: 0202432
Available in various thread sizes and materials
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Idex MicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA
Idex MicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA
0202432
₹2,400.00INR / Each
+ =
Total Price 3040.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • Connects with P-720 and P-771 unions
  • Max pressure 1000 psi (345 bar)
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Year Warranty
Specifications
  • MaterialDelrin® acetal
  • ColorNatural
  • Compatible With6-32 Coned Port
  • DescriptionMicroTight Guage Plug, Acetal, 6-32 Coned; 1/EA