Idex MicroTight Adapter, True ZDV, PEEK, 1/16" OD to MicroTight Sleeve, 10-32 to 6-32
Representative image only.
Idex MicroTight Adapter, True ZDV, PEEK, 1/16" OD to MicroTight Sleeve, 10-32 to 6-32
Idex | Mfr: P-770| Item: 0201442
Easily connect capillary tubing with tubing sleeves
₹13,400.00INR / Each
₹13,400.00INR / Each
Product Description
Includes
One F-120, one F-125, one P-554
Key Features
  • Securely join together fluid pathway components
  • Holds tight connection even through vibration
  • Frit volume 0.7 µL
  • 1 month warranty
  • 1 month warranty
More About this Item
Use tubing sleeves to connect 1/16" OD tubing to capillary tubing with IDs raging from 70 to 520 µm. MicroTight tubing sleeves have 0.025" OD. Port 1 is 10-32 coned for 1/16" OD tubing.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 2) (InchShort)6-32 Coned
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Volume (µL)0
  • ColorNatural
  • Max Operating Pressure (PSI)4000
  • DescriptionMicroTight Adapter Assembly, True ZDV, PEEK, 1/16" OD Tubing to MicroTight Sleeve, 10-32 Coned to 6-32 Coned; 1/EA