Idex MicroTight Adapter Assy, True ZDV, Blue PEEK, 1/16" to 360 µm OD, 10-32 to 6-32
Representative image only.
Idex MicroTight Adapter Assy, True ZDV, Blue PEEK, 1/16" to 360 µm OD, 10-32 to 6-32
Idex | Mfr: P-882| Item: 0201460
Create a high-pressure connection between 1/16" OD tubing and capillary tubing
₹13,400.00INR / Each
₹13,400.00INR / Each
Product Description
Includes
One F-120B, one F-124S, one P-554
Key Features
  • Securely join together fluid pathway components
  • Manufactured from 316 stainless steel or inert polymers
  • Holds tight connection even through vibration
  • Frit volume 0.6 µL
  • 1 month warranty
  • 1 month warranty
More About this Item
Port 1 is 10-32 coned for 1/16" OD tubing.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 2) (InchShort)6-32 Coned
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 2) (mm)0.36
  • Volume (µL)0
  • ColorBlue
  • Max Operating Pressure (PSI)5000
  • DescriptionMicroTight Adapter Assembly, True ZDV, Blue PEEK, 1/16" to 360 µm OD Tubing, 10-32 Coned to 6-32 Coned; 1/EA