Idex Low Pressure Chromatography Fittings Kit
Representative image only.
Idex Low Pressure Chromatography Fittings Kit
Idex | Mfr: 1456| Item: 0200852
Save time and money with a fitting kits
₹154,000.00INR / Set of 1
₹154,000.00INR / Set of 1
Product Description
Key Features
  • Constructed of chemically inert PEEK plastic
  • 1 month warranty
More About this Item
PEEK Low-Pressure Unions, Tees, and Crosses hold up to 1000 psi (1/16" tubing), or to 500 psi (1/8" tubing). PEEK flangeless fittings included. Models 02008-00, -10, and -20 take 1/16" OD tubing; models 02008-04, -14, and -24 take 1/8" OD tubing.
PEEK Fitting Kit contains convenient 1⁄4-28 threaded fittings for both 1⁄16" and 1⁄8" tubing:
1⁄16" tubing: PEEK flangeless nuts (6), Tefzel® flangeless ferrules (10), and PEEK 0.020" bore
Tee (1), cross (1), and union (2). Also includes 10-32 to M6 adapters (2) and 10-32 to 1⁄4-28
female adapters (2).
1⁄8" tubing: PEEK flangeless nuts (6), Tefzel flangeless ferrules (10), and PEEK 0.050" bore
Tee (1), cross (1), and union (2). Also includes 10-32 to M6 adapters (2) and 10-32 to 1⁄4-28
female adapters (2).
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • DescriptionLow Pressure Chromatorgraphy Fittings Kit
  • Warranty1 month