Idex LiteTouch® MicroFerrule, Natural PEEK/SS, 1/32" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex LiteTouch® MicroFerrule, Natural PEEK/SS, 1/32" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: LT-132X| Item: 0201353
High pressure fittings with fingertight convenience
₹29,900.00INR / Pkg of 10
₹29,900.00INR / Pkg of 10
Product Description
Key Features
  • Grips tubing at two pressure points
  • Design prevents tubing from twisting
  • MicroFerrule with lock ring
  • For 1/32" OD tubing
  • Max pressure 5000 psi (345 bar)
  • 1 month warranty
More About this Item
Combine a LiteTouch® nut and LiteTouch ferrule to create your fitting assembly. The LiteTouch fitting assembly grips tubing at two compression points allowing fittings to withstand high pressures with fingertight convenience. Ferrule with lock ring prevents polymer tubing from twisting. Use with hard tubing, such as stainless steel and PEEK tubing.
Fittings are not recommended for repeated use in plastic ports.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK/SS
  • Locking Sleeve MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/32
  • Compression Size / Tube Od (Port 1) (mm)0.794
  • ColorNatural
  • Max Operating Pressure (PSI)5000
  • DescriptionLiteTouch® MicroFerrule, Natural PEEK/SS, 1/32" OD Tubing, 10-32 Coned; 10/PK