Idex LiteTouch® Micro Ferrule, 1/32"; 1/EA
Representative image only.
Idex LiteTouch® Micro Ferrule, 1/32"; 1/EA
Idex | Mfr: LT-132| Item: 0202252
High pressure fittings with fingertight convenience
₹3,400.00INR / Each
₹3,400.00INR / Each
Product Description
Key Features
  • Grips tubing at two pressure points
  • Design prevents tubing from twisting
  • Applications: Amines, drugs, esters, hydrocarbons, pesticides, PONA
  • 1 year warranty
More About this Item
Combine a LiteTouch® nut and LiteTouch ferrule to create your fitting assembly. The LiteTouch fitting assembly grips tubing at two compression points allowing fittings to withstand high pressures with fingertight convenience. Ferrule with lock ring prevents polymer tubing from twisting. Use with hard tubing, such as stainless steel and PEEK tubing.
Fittings are not recommended for repeated use in plastic ports.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK/SS
  • Locking Sleeve MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/32
  • Compression Size / Tube Od (Port 1) (mm)0.794
  • ColorNatural
  • Max Operating Pressure (PSI)5000
  • DescriptionMicroFerrule, Natural PEEK/SS, 1/32" OD Tubing, 10-32 Coned; 1/EA