Idex High-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA
Representative image only.
Idex High-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA
Idex | Mfr: P-555| Item: 0202434
Available in various thread sizes and materials
₹2,600.00INR / Each
₹2,600.00INR / Each
Buy It Together
Current Item
Idex High-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA
Idex High-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA
0202434
₹2,600.00INR / Each
+ =
Total Price 3240.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • Max pressure 5000 psi (345 bar)
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorNatural
  • Compatible With6-32 Coned Port
  • DescriptionHigh-Pressure MicroTight Plug, PEEK, 6-32 Coned; 1/EA