Idex Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK
Idex | Mfr: P-225YX| Item: 0201966
Eliminate the need to flange tubing
₹13,000.00INR / Pkg of 10
₹13,000.00INR / Pkg of 10
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK
Idex Flangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK
0201966
₹13,000.00INR / Pkg of 10
+ =
Total Price 13490.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Removable and reusable—saving you money!
  • Yellow ETFE
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorYellow
  • DescriptionFlangeless Male Nut, Standard Knurl, Yellow ETFE, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK