Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Idex | Mfr: P-345X| Item: 0202038
Eliminate the need to flange tubing
₹8,100.00INR / Pkg of 10
₹8,100.00INR / Pkg of 10
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
0202038
₹8,100.00INR / Pkg of 10
+
Idex Lock Nut for 1/4-28 Flangeless Fittings, White Acetal; 10/PK
0201997
₹9,800.00INR / Pkg of 10
+ + + =
Total Price 33170.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Removable and reusable—saving you money!
  • Natural PFA
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPFA
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • DescriptionFlangeless Male Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK