Idex Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Idex | Mfr: P-226| Item: 0201967
Eliminate the need to flange tubing
₹1,600.00INR / Each
₹1,600.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
0201967
₹1,600.00INR / Each
+ =
Total Price 2090.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Removable and reusable—saving you money!
  • Black PFA
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPFA
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlack
  • DescriptionFlangeless Male Nut, Standard Knurl, Black PFA, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA