Idex Flangeless Male Nut, Standard Knurl, Black PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Black PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-230BLK| Item: 0201969
Eliminate the need to flange tubing
₹1,300.00INR / Each
₹1,300.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Black PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Standard Knurl, Black PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
0201969
₹1,300.00INR / Each
+ + =
Total Price 2750.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Removable and reusable—saving you money!
  • Black PEEK
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorBlack
  • DescriptionFlangeless Male Nut, Standard Knurl, Black PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA