Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
Idex | Mfr: P-307| Item: 0201992
Use with multiple size tubing ODs
₹510.00INR / Each
₹510.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA
0201992
₹510.00INR / Each
+ =
Total Price 1000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Short head available for use in tight-space applications
  • Standard knurl
  • Black acetal
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Ferrules are available in various materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)1.8
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlack
  • DescriptionFlangeless Male Nut, Standard Knurl, Black Acetal, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 1/EA