Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk
Idex | Mfr: P-207X| Item: 0201946
Eliminate the need to flange tubing
₹4,800.00INR / Pkg of 10
₹4,800.00INR / Pkg of 10
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk
Idex Flangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk
0201946
₹4,800.00INR / Pkg of 10
+ =
Total Price 5290.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Removable and reusable—saving you money!
  • Black acetal
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlack
  • DescriptionFlangeless Male Nut, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/Pk