Idex Flangeless Male Nut, Standard Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Standard Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-218| Item: 0201956
Short head is ideal for tight-space applications
₹760.00INR / Each
₹760.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Standard Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Standard Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
0201956
₹760.00INR / Each
+ + =
Total Price 18540.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural ETFE
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16