Idex Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
Idex | Mfr: P-347BX| Item: 0202041
Use with multiple size tubing ODs
₹14,300.00INR / Pkg of 10
₹14,300.00INR / Pkg of 10
Buy It Together
Current Item
Idex Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
Idex Flangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
0202041
₹14,300.00INR / Pkg of 10
+ =
Total Price 19000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Short head available for use in tight-space applications
  • Short head
  • Blue PEEK
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Ferrules are available in various materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)1.8
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlue
  • DescriptionFlangeless Male Nut, Short Head, Blue PEEK, 1.8, 2.0, 3.0 mm, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK