Idex Flangeless Fitting, Large Knurl, Black PEEK, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Fitting, Large Knurl, Black PEEK, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Idex | Mfr: XU-662| Item: 0202105
Eliminate the need to flange tubing
₹3,300.00INR / Each
₹3,300.00INR / Each
Product Description
Includes
Natural ETFE ferrule 02020-50.
Key Features
  • Removable and reusable—saving you money!
  • Black PEEK
  • For 5/16" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/2-20 flat bottom
  • Compression Size / Tube Od (Port 1) (in)5/16
  • ColorBlack
  • DescriptionFlangeless Fitting, Large Knurl, Black PEEK, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 1/EA