Idex Flangeless Fitting Kit, Acetal Nuts and ETFE Ferrules, 1/16" and 1/8" OD Tubing; 1/EA
Representative image only.
Idex Flangeless Fitting Kit, Acetal Nuts and ETFE Ferrules, 1/16" and 1/8" OD Tubing; 1/EA
Idex | Mfr: 1451| Item: 0201920
Assortment of flangeless fittings to meet you everyday needs
₹102,000.00INR / Each
₹102,000.00INR / Each
Buy It Together
Current Item
Idex Flangeless Fitting Kit, Acetal Nuts and ETFE Ferrules, 1/16" and 1/8" OD Tubing; 1/EA
Idex Flangeless Fitting Kit, Acetal Nuts and ETFE Ferrules, 1/16" and 1/8" OD Tubing; 1/EA
0201920
₹102,000.00INR / Each
+ =
Total Price 102490.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • For 1/16" and 1/8" OD tubing
  • 1/4-28 flat-bottom connection
  • 1 year warranty
More About this Item
Various colors available for 1/16" OD and 1/8" OD tubing.
Kit contains:
For 1/16" OD tubing:black (10), blue (10), cream (10), green (10), red (10), and white (10) Delrin® acetal nuts and blue ETFE ferrules (60).
For 1/8" OD tubing:black (10), blue (10), cream (10), green (10), red (10), and white (10) Delrin® acetal nuts and yellow ETFE ferrules (60).
1 Year Warranty
Specifications
  • Fitting MaterialDelrin® acetal
  • Thread size UNF(M) (port 1) (InchShort)1/4-28 flat bottom
  • DescriptionFlangeless Fitting Kit, Acetal Nuts and ETFE Ferrules, 1/16" and 1/8" OD Tubing; 1/EA