Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-582B| Item: 0202579
Excellent for low to moderate pressure applications
₹1,300.00INR / Each
₹1,300.00INR / Each
Buy It Together
Current Item
Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex Flanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
0202579
₹1,300.00INR / Each
+ =
Total Price 2270.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Stainless steel washer.
Stainless steel washer.
Key Features
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • For 1/8" OD tubing
  • Blue Delrin® acetal nut
  • 1 year warranty
More About this Item
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
Use with flanged tubing. Delrin acetal nuts resist cross threading or loosening during use.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorBlue
  • DescriptionFlanged Fitting, Standard Knurl, Blue Acetal, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA