Idex Flanged Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Flanged Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Idex | Mfr: P-982BLKX| Item: 0202581
Excellent for low to moderate pressure applications
₹7,100.00INR / Pkg of 10
₹7,100.00INR / Pkg of 10
Product Description
Includes
Stainless steel washer.
Key Features
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • 1 year warranty
More About this Item
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 1) (mm)6
  • ColorBlack
  • DescriptionFlanged Fitting, Standard Knurl, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA