Idex Flanged Fitting Kit, 1/16" and 1/8" OD Tubing, 1/4-28 Flat-Bottom
Representative image only.
Idex Flanged Fitting Kit, 1/16" and 1/8" OD Tubing, 1/4-28 Flat-Bottom
Idex | Mfr: 1453| Item: 0202552
Excellent for low to moderate pressure applications
₹125,000.00INR / Each
₹125,000.00INR / Each
Product Description
Key Features
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • Delrin® acetal nuts and SS washers
  • 1 year warranty
More About this Item
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)1/4-28 flat bottom
  • ColorBlack, red, white, cream, green, and blue
  • DescriptionFlanged Fitting Kit, 1/16" and 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA