Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA
Representative image only.
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA
Idex | Mfr: F-330| Item: 0201910
Popular threaded coned port configurations support quality high-pressure connections
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA
Idex Fingertight Two-Piece Fitting, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA
0201910
₹2,400.00INR / Each
+ =
Total Price 4000.00 ADD 2 ITEMS TO CART
Product Description
Includes
PEEK fittings include PEEK ferrule 02007-50; SS fittings include PEEK Ferrule 02007-52.
Key Features
  • Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
  • Applications: Amines, drugs, esters, hydrocarbons, pesticides, PONA
  • 1 year warranty
More About this Item
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned, long thread
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)6000
  • DescriptionFingertight Two-Piece, Long Thread, Natural PEEK, 1/16" OD Tubing, 10-32 Coned, 1/EA