Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: F-300X| Item: 0201347
Withstands up to 6000 psi!
₹22,500.00INR / Pkg of 10
₹22,500.00INR / Pkg of 10
Buy It Together
Current Item
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK
0201347
₹22,500.00INR / Pkg of 10
+ =
Total Price 24100.00 ADD 2 ITEMS TO CART
Product Description
Includes
Natural PEEK ferrule 02007-50.
Key Features
  • Winged nut head provides extra leverage when tightening into a receiving port
  • Double wing
  • 10-32 coned
  • 10/PK
  • 1 month warranty
More About this Item
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)6000
  • DescriptionF-300X Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 10/PK