Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Representative image only.
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Idex | Mfr: F-300| Item: 0200700
Withstands up to 6000 psi!
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA
0200700
₹2,400.00INR / Each
+ =
Total Price 4000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Natural PEEK ferrule 02007-50.
Key Features
  • Winged nut head provides extra leverage when tightening into a receiving port
  • Double wing
  • 10-32 coned
  • 1/EA
More About this Item
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)6000
  • DescriptionFingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA