Idex Fingertight One-Piece Fitting, Diamond Knurl, Natural PCTFE, 1/8" OD Tubing, 1/4-28 Coned; 1/EA
Representative image only.
Idex Fingertight One-Piece Fitting, Diamond Knurl, Natural PCTFE, 1/8" OD Tubing, 1/4-28 Coned; 1/EA
Idex | Mfr: P-100| Item: 0201918
Diamond knurl provides an extra grip for tightening the fitting
₹4,100.00INR / Each
₹4,100.00INR / Each
Product Description
Key Features
  • Constructed of natural PCTFE
  • Applications: Amines, drugs, esters, hydrocarbons, pesticides, PONA
  • 1 year warranty
More About this Item
Get the convenience of nut and ferrule built into one fitting. By using one-piece fittings there is no need to worry about the ferrule getting stuck in a receiving port.
1 Year Warranty
Specifications
  • Fitting MaterialPCTFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 coned
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • Max Operating Pressure (PSI)1000
  • DescriptionFingertight One-Piece, Diamond Knurl, Natural PCTFE, 1/8" OD Tubing, 1/4-28 Coned; 1/EA