Idex Fingertight Ferrule, PEEK, 1/32" OD Tubing, 10-32 Coned; 1/EA
Representative image only.
Idex Fingertight Ferrule, PEEK, 1/32" OD Tubing, 10-32 Coned; 1/EA
Idex | Mfr: F-113| Item: 6300430
Features excellent chemical resistance and superior thread strength
₹4,400.00INR / Each
₹4,400.00INR / Each
Product Description
Key Features
  • Optimize performance of your IDEX Two-Piece Fingertight Fittings
  • For 1/32" OD tubing
More About this Item
Designed for use with two-piece fittings. Choose from a wide range of ferreles with a wide range of materials.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/32
  • Compression Size / Tube Od (Port 1) (mm)0.8
  • ColorNatural
  • Max Operating Pressure (PSI)6000
  • DescriptionFingertight Ferrule, PEEK, 1/32" OD Tubing, 10-32 Coned; 1/EA