Idex Ferrules for TitanEX Valve, 1/8" OD Tubing; 25/PK
Representative image only.
Idex Ferrules for TitanEX Valve, 1/8" OD Tubing; 25/PK
Idex | Mfr: 7770-039| Item: 0202523
Optimize performance of your Idex® valve
₹45,700.00INR / Pkg of 25
₹45,700.00INR / Pkg of 25
Product Description
Key Features
  • Choose from replacement fittings and o-rings to extend the life of your valves
  • Use with 1/8" OD Tubing, 25/PK
  • 1 year warranty
More About this Item
Replacement ferrules and O-rings for valves offer a tight fit for 1/16" and 1/8" OD tubing.
1 Year Warranty
Specifications
  • Compatible WithTitanEX valve
  • DescriptionIdex Ferrules for TitanEX Valve, 1/8" OD Tubing; 25/PK