Idex Ferrule, Stainless Steel, 1/8" OD Tubing, 1/4-28 UNF Coned; 1/EA
Representative image only.
Idex Ferrule, Stainless Steel, 1/8" OD Tubing, 1/4-28 UNF Coned; 1/EA
Idex | Mfr: C-236| Item: 4271849
Can withstand pressures up to 20,000 psi
₹2,200.00INR / Each
₹2,200.00INR / Each
Product Description
Key Features
  • Durable stainless steel
  • Great for connecting tubing
  • 1 year warranty
More About this Item
This ferrule is about strength and reliability. When tightened with a nut, the stainless steel ferrule can withstand pressures up to 20,000 psi.
1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Max Operating Pressure (PSI)20000
  • DescriptionFerrule, SS, 1/8" OD Tubing, 1/4-28 UNF; 1/EA