Idex Coupler, PEEK, Male/Male-1/2"
Representative image only.
Idex Coupler, PEEK, Male/Male-1/2"
Idex | Mfr: U-288| Item: 4271940
Great for reducing overall flow path volume
₹19,700.00INR / Each
₹19,700.00INR / Each
Product Description
Key Features
  • Couple two devices together
  • Lessen volume of flow path
  • 10-32 coned port connections
  • Applications: Alcohols, aromatics, flavors, free fatty acids, low-polarity pesticides
  • 1 year warranty
More About this Item
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
This Column Coupler delivers a low swept-volume connection, which is an excellent option for reducing overall flow path volume, while coupling together two devices. It is manufactured from biocompatible PEEK, which has an excellent chemical resistance to virtually all commonly used solvents.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorNatural
  • DescriptionColumn Coupler, PEEK, Male/Male-1/2," 10-32 Coned; 1/EA