Idex Column Coupler; PCTFE, 10-32 Coned; 1/EA
Representative image only.
Idex Column Coupler; PCTFE, 10-32 Coned; 1/EA
Idex | Mfr: U-283| Item: 0193995
Great for reducing overall flow path volume
₹17,900.00INR / Each
₹17,900.00INR / Each
Product Description
Includes
Two diamond knurl PCTFE, one-piece fingertight fitting 02018-71.
Key Features
  • Couple two devices together
  • Lessen volume of flow path
  • 10-32 coned port connections
  • One-piece PCTFE nut/ferrule
  • Max pressure rating of 4000 psi (276 bar)
More About this Item
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
Specifications
  • MaterialPCTFE
  • ColorRed
  • DescriptionColumn Coupler, PCTFE, 10-32 Coned; 1/EA