Idex Column Coupler, Acetal/PEEK, 10-32 Coned; 1/EA
Representative image only.
Idex Column Coupler, Acetal/PEEK, 10-32 Coned; 1/EA
Idex | Mfr: U-284| Item: 0193996
Great for reducing overall flow path volume
₹6,800.00INR / Each
₹6,800.00INR / Each
Product Description
Includes
Two single-wing acetal, two-piece fingertight fitting 02019-05
Key Features
  • Couple two devices together
  • Lessen volume of flow path
  • 10-32 coned port connections
  • 0.007" thru-hole
  • 1.3 µL swept volume
  • Acetal nut/PEEK ferrule
  • Max pressure of 5000 psi (345 bar)
More About this Item
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
Specifications
  • MaterialAcetal nut/PEEK ferrule
  • ColorRed
  • DescriptionColumn Coupler, Acetal/PEEK, 10-32 Coned; 1/EA