Idex Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA

Representative image only.

Idex Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA

Idex | Mfr: A-350| Item: 0201330

Easily make clean cuts with tubing cutters

₹54,000.00INR / Each

₹54,000.00INR / Each

Buy It Together

Current Item

Idex Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA
Idex Capillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA
0201330

₹54,000.00INR / Each
+ + +
Idex A-328 Replacement Blades for Polymer Tubing Cutters; 5/PK
0201686

₹2,500.00INR / Pkg of 5
+ + =
Total Price 100800.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Ten 2" long FEP tubing sleeves for 360 µm OD tubing and five replacement blades.

Key Features

  • Cutters are available for different tubing sizes and materials
  • For use with 360 µm to 1/32" OD polymer tubing
  • 1 month warranty

More About this Item

Choose from tubing cutters for a wide range of tubing sizes, suitable for a wide range of applications.

Capillary polymer tubing cutter makes clean, perpendicular cuts without collapsing thin capillary walls. Order tubing sleeves for 1/32" OD separately.

1 Month Warranty
Specifications
  • For Tubing OD (in)1/64
  • Max Tubing OD (in)1/32
  • DescriptionCapillary Polymer Chromatography Tubing Cutter For 360 µm to 1/32" OD tubing; 1/EA
Reviews
Q&A