Idex Barbed Connector, Straight, Polypropylene, 3/8" x 3/8" Hose Barb
Representative image only.
Idex Barbed Connector, Straight, Polypropylene, 3/8" x 3/8" Hose Barb
Idex | Mfr: P-805| Item: 0202388
Make solid connections in low-pressure applications
₹450.00INR / Each
₹450.00INR / Each
Product Description
Key Features
  • Manufactured from polypropylene
  • For use with 3/8" ID soft-walled tubing
  • 1 year warranty
More About this Item
Use barbed straight connectors to firmly join together two pieces of soft-walled peristaltic tubing with similar inner diameters.
1 Year Warranty
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeHPLC/Chromatography
  • Barb Size / Tubing Id (Port 1) (mm)9.5
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • Barb Size / Tubing Id (Port 2) (mm)9.5
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • ColorNatural
  • DescriptionLow-Pressure Connector, Barbed, Polypropylene, 3/8 x 3/8" ID Tubing; 1/EA