Idex Barbed Connector, Straight, Polypropylene, 3/16" x 3/16" Hose Barb; 1/EA
Representative image only.
Idex Barbed Connector, Straight, Polypropylene, 3/16" x 3/16" Hose Barb; 1/EA
Idex | Mfr: P-803| Item: 0202387
Make solid connections in low-pressure applications
₹220.00INR / Each
₹220.00INR / Each
Product Description
Key Features
  • Manufactured from polypropylene
  • For use with 3/16" ID soft-walled tubing
  • 1 year warranty
More About this Item
Use barbed straight connectors to firmly join together two pieces of soft-walled peristaltic tubing with similar inner diameters.
1 Year Warranty
Specifications
  • Fitting MaterialPolypropylene
  • Connection TypeHPLC/Chromatography
  • Barb Size / Tubing Id (Port 1) (mm)4.8
  • Barb Size / Tubing Id (Port 1) (in)3/16
  • Barb Size / Tubing Id (Port 2) (mm)4.8
  • Barb Size / Tubing Id (Port 2) (in)3/16
  • ColorNatural
  • DescriptionLow-Pressure Connector, Barbed, Polypropylene, 3/16 x 3/16" ID Tubing; 1/EA