Idex Back Pressure Regulator Cartridge, 750 psi; 1/EA

Representative image only.

Idex Back Pressure Regulator Cartridge, 750 psi; 1/EA

Idex | Mfr: P-795| Item: 0201726

Select from preset pressure settings

₹30,300.00INR / Each

₹30,300.00INR / Each
Product Description

Key Features

  • Use with BPR holders
  • Blue end cap, black body
  • 1 year warranty

More About this Item

Use BPR cartridges with the holders to enhance system performance through outgassing prevention and improved pump check valve efficiency.

1 Year Warranty
Specifications
  • Connection TypeHPLC/Chromatography
  • Pressure Setting (PSI)750
  • DescriptionIdex Back Pressure Regulator Cartridge, 750 psi; 1/EA
  • Warranty1 year
Reviews
Q&A