Ibi Scientific Histochemistry Staining Tray, 12-place, black lid

Representative image only.

Ibi Scientific Histochemistry Staining Tray, 12-place, black lid

Ibi Scientific | Mfr: AT-12| Item: 0048582

Designed specifically for immunohistochemistry slide staining

₹68,000.00INR / Each

₹68,000.00INR / Each
Product Description

Includes

lid.

More About this Item

These staining trays work well in microscopy, cytology, and hematology settings and are designed for immunohistochemistry assay slide staining in the lab. All trays have an integrated dual-well design reservoir to hold water beneath the slides when humidity is needed. The skid-proof rubber feet prevent sliding and allow for stable stacking for a tidy benchtop.

Made from 100% heavy-duty black cast acrylic for years of robust use, while also blocking all light from the stained slides.

Specifications
  • Capacity (Number of slides)12
  • External Width (in)7.5
  • External Height (in)1
  • External Depth (in)10 1/2
  • External Width (cm)19.05
  • External Height (cm)2.54
  • External Depth (cm)26.67
  • DescriptionHistochemistry Staining Tray, 12-place, black lid
Reviews
Q&A