Hydrion Spill Sticks PH Spill Stick, Range 0-13, 10/pk
Representative image only.
Hydrion Spill Sticks PH Spill Stick, Range 0-13, 10/pk
Hydrion | Mfr: DS-8020| Item: 5920005
Keep a safe distance with pH spill sticks
₹4,700.00INR / Each
₹4,700.00INR / Each
Product Description
More About this Item
Extra-long 6” (15.2 cm) pH sticks help reduce the risk of contact with the sample—ideal for chemical spills or any materials you want to avoid touching. Sticks give a simple, single color match for all pH values from 0 to 13 in distinct, bright color separations. Includes ten pH spill sticks and color matching chart.
Pack of 10 strips and color chart.
Specifications
  • Test pH
  • Measurement Points0 to 13 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit10
  • DescriptionPH Spill Stick, Range 0-13, 10/pk