Hydrion PAA160 Peracetic Acid (Paa) Sanitizer Test Strips, 50 Strips/Vial; 6 Vials/Cs
Representative image only.
Hydrion PAA160 Peracetic Acid (Paa) Sanitizer Test Strips, 50 Strips/Vial; 6 Vials/Cs
Hydrion | Mfr: PAA160| Item: 5920055
Quick reliable test for the strength of PPA wash solutions
₹20,500.00INR / Pkg of 6
₹20,500.00INR / Pkg of 6
Product Description
Key Features
  • Covers critical ranges for antimicrobial fruit and vegtable rinses; 0 to 160 ppm
  • Meets federal, state, and local food safety guidelines
More About this Item
Simple test for the strength and efficacy of PAA antimicrobial vegtable and fruit wash solutions in food service. Just dip the test strip and match to scale on the flip-top vial within ten seconds. Packaged as a carton of 6 vials with 50 test strips per vial.
Specifications
  • TestParaacetic acid (PAA)
  • Measurement Points0 to 160 ppm
  • Reagent PlatformStrips
  • Tests Per Kit300
  • DescriptionPeracetic Acid (Paa) Sanitizer Test Strips, 50 Strips/Vial; 6 Vials/Cs