Hydrion P-12M Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
Representative image only.
Hydrion P-12M Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
Hydrion | Mfr: P-12M| Item: 5920032
Immediately gives the surface reaction of any solid to the nearest pH value
₹9,600.00INR / Pkg of 3
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹9,600.00INR / Pkg of 3
Buy It Together
Current Item
Hydrion P-12M Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
Hydrion P-12M Insta-Chek Mechanical pH Pencil, pH 0-13, 3 per package
5920032
₹9,600.00INR / Pkg of 3
+ =
Total Price 34300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Retractable, nonrefillable, mechanical pencils are more convenient than paper-wrap pencils
  • The widest range available — reading from pH 0 to 13 — with 14 different color matches
  • 2 year warranty
More About this Item
The handy, easily portable form is perfect for pH spot checks, requiring as little as 0.001 mL of buffered solution. Use pencil on virtually any surface—even irregular or uneven ones. It is effective on porous material and impervious solids.
To use, first wet the surface or area with distilled water and stroke several times with the pH pencil. After 15 seconds, compare the resulting color against the color chart. Pack includes three mechanical pencils and two color charts.
Two year warranty
Specifications
  • TestpH
  • Measurement Points0 to 13 pH in 1 pH unit increments
  • DescriptionInsta-Chek Mechanical pH Pencil, pH 0-13, 3 per package