Hydrion IM-220 iodine test paper, 15-ft roll, box of 10
Representative image only.
Hydrion IM-220 iodine test paper, 15-ft roll, box of 10
Hydrion | Mfr: IM-220| Item: 5920054
Convenient dispenser roll lets you use the exact strip length needed
₹13,200.00INR / Box of 10
₹13,200.00INR / Box of 10
Product Description
Key Features
  • Reliable and economical means to measure sanitizer strength
  • 1 year warranty
More About this Item
Federal, state, and local health regulations require the use of test kits to verify the strength of sanitizer solutions. Hydrion test strips are a quick, reliable, and economical solution for this. Test strips are available to test quaternary ammonium, chlorine, and iodine-based sanitizers. Strips provide a visual color-change indication of solution strength. Each 15-ft (4.6-m) roll comes with a color match chart and is clearly labeled. Each roll provides enough paper for about 100 tests.
1 Year Warranty
Specifications
  • TestIodine
  • Measurement Points50, 75, 150, 225 ppm
  • Tests Per Kit10
  • DescriptionHydrion iodine test paper, 15-ft roll, box of 10