Hydrion 3.0 to 5.5 pH Jumbo pH Papers, Range 3.0 to 5.5, 50 Ft/Roll
Representative image only.
Hydrion 3.0 to 5.5 pH Jumbo pH Papers, Range 3.0 to 5.5, 50 Ft/Roll
Hydrion | Mfr: HJ-626| Item: 5920026
Larger, field-sized papers for quick pH monitoring
₹3,800.00INR / Carton of 1
₹3,800.00INR / Carton of 1
Product Description
Includes
1 roll of paper and protective case.
More About this Item
These low-cost jumbo rolls offer larger paper strips to measure pH in one of four ranges. Ideal for use in the field or plant. The larger size makes it easier for testing--even with gloves.
Specifications
  • TestpH
  • Measurement Points3.0 to 5.5 pH in 0.5 pH unit increments
  • DescriptionJumbo pH Papers, Range 3.0 to 5.5, 50 Ft/Roll