HTL 7903 Discovery Pro Starter Kit with DP10, DP20, DP200 & DP1000 Pipettors and Accessories
Representative image only.
HTL 7903 Discovery Pro Starter Kit with DP10, DP20, DP200 & DP1000 Pipettors and Accessories
| Mfr: 7903| Item: 2455219
Go "Pro" to reduce strain and ward off pain
Call For Price
Call For Price
Product Description
Includes
pipettes, stand, 10 uL, 200 uL, and 1200 uL tips, calibration tool, and pipette hanger.
Key Features
  • Ultra-lightweight and ergonomic handle design; correct grip is ensured
  • Tip ejection force is considerably lower than traditional pipettes
  • Compatible with virtually all brands of tips
  • Fully autoclavable
  • Large 4-digit counter is easily visible
  • 3 year warranty
More About this Item
Don't stress! Along with optimal precision and accuracy, the Discovery Pro adjustable-volume pipettors are designed to maximize user comfort. Made from the high-quality grade materials, innovative features include a contoured handle, unique pushbutton-lever system, finger rest and a 4-digit counter that rewinds quickly. Additionally, it's a snap to set volume--and lock it in--while the pipettor's smart design ensures thermal insulation of internal parts, improving accuracy and durability.
3 Year Warranty
Specifications
  • AutoclavableYes
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed, Yellow, and Blue
  • DescriptionStarter Kit with DP10, DP20, DP200 & DP1000 Pipettors and Accessories