HTL 6001 Discovery Pro DP2 Single Channel Pipette, Red; 0.2 to 2 µL
Representative image only.
HTL 6001 Discovery Pro DP2 Single Channel Pipette, Red; 0.2 to 2 µL
| Mfr: 6001| Item: 2455203
Go "Pro" to reduce strain and ward off pain
Call For Price
Call For Price
Product Description
Key Features
  • Ultra-lightweight and ergonomic handle design; correct grip is ensured
  • Tip ejection force is considerably lower than traditional pipettes
  • Compatible with virtually all brands of tips
  • Fully autoclavable
  • Large 4-digit counter is easily visible
  • 3 year warranty
More About this Item
Don't stress! Along with optimal precision and accuracy, the Discovery Pro adjustable-volume pipettors are designed to maximize user comfort. Made from the high-quality grade materials, innovative features include a contoured handle, unique pushbutton-lever system, finger rest and a 4-digit counter that rewinds quickly. Additionally, it's a snap to set volume--and lock it in--while the pipettor's smart design ensures thermal insulation of internal parts, improving accuracy and durability.
3 Year Warranty
Specifications
  • Max Volume (µL)2
  • AutoclavableYes
  • Min Volume (µL)0.2
  • Accuracy (%)±1.5
  • Precision (%)±0.7
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeRed
  • DescriptionSingle Channel Pipette, Red; 0.2 to 2 µL