HTL 5147 Optipette OP12-200 Multichannel Pipettor, Yellow, 12-Channel; 20 to 200 µL
Representative image only.
HTL 5147 Optipette OP12-200 Multichannel Pipettor, Yellow, 12-Channel; 20 to 200 µL
| Mfr: 5147| Item: 2455177
Easily adjust ejector height
Call For Price
Call For Price
Product Description
Key Features
  • Ergonomic handle design
  • Easy to calibrate
  • Rotating pipetting push button prevents accidental volume change
  • 3 year warranty
More About this Item
Remarkably robust and durable in a sleek design, Optipette adjustable-volume pipettors offer optimal accuracy and precision. With a UV-resistant body and autoclavable bottom half, the soft spring system reduces pipetting forces while color-coded rings simplify volume identification. Additional spacers enable ejector height adjustment; user-calibration is easy thanks to a provided calibration key.
3 Year Warranty
Specifications
  • Max Volume (µL)200
  • AutoclavableYes
  • Min Volume (µL)20
  • Accuracy (%)±1
  • Precision (%)±0.6
  • Number Of Channels12
  • Tip EjectorYes
  • Color CodeYellow
  • DescriptionMultichannel Pipettor, Yellow, 12-Channel; 20 to 200 µL